MAPA® Unlined Z Pattern Nitrile Gloves, Magid
Supplier: Magid Glove
A158 M157
32889-304CS 606.74 CAD
32889-304 32889-306 32889-392
MAPA® Unlined Z Pattern Nitrile Gloves, Magid
Gloves
Nitrile glove featuring high puncture resistance.
  • 100% nitrile
  • Embossed grip
  • unisex gloves
They also feature good solvent and chemical resistant qualities.