Magid® CleanMaster® Lightweight Nylon Gloves, Magid
Supplier: Magid Glove
4511S 4511L
32933-908PK 140.97 CAD
32933-908 32933-912
Magid® CleanMaster® Lightweight Nylon Gloves, Magid
Gloves
The Magid CleanMaster nylon tricot woman's cleanroom gloves are designed to specifically fit a woman's hand.
  • Two-piece nylon tricot glove
  • Cut and sewn
  • Low linting for less contamination
  • Color-coded hem for easy matching
These gloves feature shorter finger lengths, a narrower palm and a repositioned thumb for better fit and dexterity.