Magid® ROC® Polyurethane Palm Coated Gloves, Magid
Supplier: Magid Glove
GP1506 GP1507 GP1508
32893-138CS 672 CAD
32893-138 69100-430 69100-432 69100-434
Magid® ROC® Polyurethane Palm Coated Gloves, Magid
Gloves
Work Gloves feature a durable, silicone free, polyurethane palm coating that provides excellent grip in either oily or dry conditions.
  • Breathable, form-fitting, 100% stretch nylon shell
These gloves with gauge, seamless, machine knit shell and knit wrist cuff, are form-fitting delivering superb comfort and breathability during even the longest shift.
The ROC® Glove's nylon shell is also tough providing good abrasion resistance and ANSI Cut Level 1 protection. Ideal choice in hand protection for general maintenance, assembly, and material handling.With the ROC, it feels like working with your bare hands.
Packaging: Packed one dozen to a polybag.