Magid® CleanMaster® Hemmed Nylon Tricot Gloves, Magid
Supplier: Magid
4312-XL 4312-L 4312-M
32889-454PK 201.37 CAD
32889-454 32889-456 32889-452
Magid® CleanMaster® Hemmed Nylon Tricot Gloves, Magid
Gloves
These gloves are comfortable and durable choice for cleanroom, paint and electronics projects. This style is hemmed, with a two-piece cut and sewn pattern.
  • Two-piece nylon tricot glove
  • Low linting for less contamination
  • Color-coded hem for easy matching
  • Reversible, extending wear
These gloves are reversible and fits either hand, ensuring longer wear and greater ease in sorting after laundry.