DuPont™ Coveralls with Elastic Wrists/Ankles, Magid Glove
Supplier: Magid Glove
CVC11L CVC11XL CVC11XXL CVC11XXXL
13450-174CS 846.14 CAD
13450-174 13450-176 13450-178 13450-180
DuPont™ Coveralls with Elastic Wrists/Ankles, Magid Glove
Disposable Garments
Zippered coveralls are made of Tyvek, a tough polyolefin material which provides the best balance of protection, durability and comfort.
  • Elastic border at wrists and ankles
  • Disposable
Greater range of movement while stretching and bending. Tailored fit and reinforcement in high stress areas. Tear and puncture resistant garments that withstand most chemicals and acids.