Magid® ComfortFlex® M13 Unlined Z Pattern Nitrile Gloves, Magid
Supplier: Magid Glove
M1310 M1311 M138 M139
69100-502CS 539.59 CAD
69100-502 69100-504 69100-506 69100-508 69100-510
Magid® ComfortFlex® M13 Unlined Z Pattern Nitrile Gloves, Magid
Gloves
Unlined 100% nitrile glove featuring high puncture resistance, an embossed grip, and excellent chemical resistance, all at a low cost.
  • M13 is tough and versatile enough to handle any job that comes their way
This style is also USDA accepted for use in food processing settings.