Vikan® 50" Basic Aluminum Handles, Remco Products
Supplier: Remco Products
29582 29583 29584 29585 29586 29587 29588 29589
76006-620EA 22.7 CAD
76006-620 76006-622 76006-624 76006-626 76006-628 76006-630 76006-632 76006-634
Vikan® 50" Basic Aluminum Handles, Remco Products
Mops
This standard broom handle can be used with all brooms, squeegees, and scrapers.
  • Durable handle
  • Hanging hole for easy storage
  • Hygienic Euro thread prevents walk-off and is easy to clean
An ideal solution for cleaning pipes, tubes, and drains, this handle is great for sanitation uses that do not require high manual pressure. When combined with any Remco brush heads, it makes an economical and efficient solution for hygiene needs.
Certifications: Manufactured from FDA compliant materials.
Ordering information: Accessories not included.