Dispenser Refill Packs for Sentino® Filter Dispenser, Cytiva (Formerly Pall Lab)
Supplier: Cytiva
68121 68121 68123 68124 68124 68125 68125 68123
CA30617-198PK 899 CAD
30617-200 CA30617-198 30617-202 CA30617-200 30617-198 30617-204 CA30617-202 CA30617-204
Dispenser Refill Packs for Sentino® Filter Dispenser, Cytiva (Formerly Pall Lab)
Filters Membrane Filters
These membranes have been designed specifically for use with the Sentino® automatic filter dispenser.
  • Ideal for high volume laboratories
  • Eliminate the time and waste of individually sealed packs of membrane
Delivery information: Each box contains two additional discs for loading the dispenser.