Vaplock™ Wako 38 mm Waste Caps and Waste Kits, Cole-Parmer
Supplier: Antyila Scientific
12018-68 12018-69 12018-70 12018-71 12018-72 12018-73
76287-756KT 564.94 CAD
76287-756 76287-758 76287-760 76287-762 76287-764 76287-766
Vaplock™ Wako 38 mm Waste Caps and Waste Kits, Cole-Parmer
Closures Safety Caps and Safety Waste Caps
The Vaplock™closed system design reduces chemical evaporation and exposure to hazardous vapors.
  • Minimize exposure to harmful chemical vapors
  • Collect solvent waste from multiple effluent streams
  • Highly adaptable and conformable to a broad range of containment types
Featuring a vapor tight connection, waste manifolds are designed to fit standard GL38 threaded lab bottles. Chemical-resistant materials of construction include polypropylene or PTFE manifold and insert, with polypropylene spin collar. These versatile manifolds can be configured for barbedMore Product Information