Vaplock™ Safety Waste Funnel, Cole-Parmer
Supplier: Antyila Scientific
12020-64 12020-65 12020-66
76287-646EA 444.18 CAD
76287-646 76287-648 76287-650
Vaplock™ Safety Waste Funnel, Cole-Parmer
Funnels Safety Funnels
Vaplock™ SF Series funnels allow for safer disposal of pourable laboratory waste.
Vaplock™ SF Series funnels allow for safer disposal of pourable laboratory waste.
As with all solvent applications, chemical compatibility should be noted prior to purchase.
A locking, hinged lid minimizes leaks and spills when the funnel is closed. If used in tandem with other Vaplock™ products, such as a manifold and exhaust filter, a closed system may be maintained for passive wastes while the funnel is sealed.