Vaplock™ GL 32 Supply Caps, Cole-Parmer
Supplier: Antyila Scientific
12018-48 12018-50 12018-51 12018-52 12018-53
76287-542EA 111.74 CAD
76287-542 76287-544 76287-546 76287-548 76287-550 76287-680
Vaplock™ GL 32 Supply Caps, Cole-Parmer
Closures Safety Caps and Safety Waste Caps
The Vaplock™ closed system design reduces chemical evaporation and exposure to hazardous vapors with Vaplock™ solvent reservoir caps.
  • Anti-twist design prevents twisting of tubing when the cap is attached or removed as the inner body rotates independently of the outer collar
  • Chemical compatibility with laboratory solvents
Durable port threads are designed to accept industry-standard 1/4-28 threaded fittings. Some caps include robust stainless steel thread inserts to minimize cross-threading and damage to soft PTFE ports. For use with semi-rigid tubingMore Product Information