Vaplock™ Tubing Connections, Cole-Parmer
Supplier: Antyila Scientific
12020-05 12020-06 12020-07 12020-08 12020-02 12020-03 12020-04
76287-576EA 133.97 CAD
76287-576 76287-578 76287-580 76287-582 76287-910 76287-912 76287-914 76287-916
Vaplock™ Tubing Connections, Cole-Parmer
Fittings and Connectors
Liquid-tight tubing connections for 1/4-28 and 5/16-24 ports.
Compression fittings designed for liquid-tight connection of semi-rigid tubing to flat-bottom 1/4-28 and 5/16-24 port threads, for 1/16" to 3/16" O.D. tubing. Each connection is comprised of a two-piece nut and ferrule system, useful for pressurized applications under 500 psi. Select based on tubing outer diameter. Fittings are color-coded by tubing size for easy identification. These bottom-sealing UNF fittings do not require PTFE tape.
Use with 1/16" to 3/16" O.D. tubing.