Brilliant III Ultra-Fast SYBR® Green ROX QPCR Master Mix, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES (GENOMICS) CA
600892
600889
600904
600903
CA76204-812EA
457.35
CAD
CA76204-812
CA76204-816
CA76204-848
CA76204-850
Brilliant III Ultra-Fast SYBR® Green ROX QPCR Master Mix, Agilent Technologies
Nucleic Acid Reagents
qPCR/RT-qPCR Enzymes and Kits
High performance, ultra-sensitive, SYBR® Green QPCR master mix reagent with ROX concentration for reliable quantification across a wide range of targets and templates.
- Reliable and reproducible data
- Novel fast Taq mutant for qPCR results in under 35 minutes Resistant to common QPCR inhibitors ( whole blood, humic acid, NaCl)
- Enhanced rapid hot start capability saves time and reduces primer-dimer formation
- Convenient pre-blended formulations compatible with any SYBR Green dye
Available with low or high ROX concentrations.