Vaplock™ Port Plugs, Cole-Parmer
Supplier: Antyila Scientific
12020-49 12020-50 12020-51 12020-44 12020-45 12020-46 12020-47 12020-48
76287-616EA 30.38 CAD
76287-616 76287-618 76287-620 76287-892 76287-894 76287-896 76287-898 76287-940
Vaplock™ Port Plugs, Cole-Parmer
Closures Chromatography Vial Closures
Plugs are intended to seal unused manifold and bottle cap ports.
Port Plugs for NPT and UNF ports are available in multiple materials, and are designed to seal unused manifold and bottle cap ports. Bottom-sealing UNF thread options include 1/4-28 ports and 5/16-24, and do not require PTFE tape. Options for NPT ports are 1/8", 1/4", 3/8", and 1/2" NPT. Plugs should be wrapped 3 to 4 times with PTFE tape for leak-resistant connections.