Vaplock™ Drum Waste Caps (2" Fine Thread) and Waste Kits, Cole-Parmer
Supplier: Antyila Scientific
12019-61 12019-62 12019-63 12019-64 12019-65 12019-66
76287-464EA 336.17 CAD
76287-464 76287-466 76287-468 76287-470 76287-472 76287-474
Vaplock™ Drum Waste Caps (2" Fine Thread) and Waste Kits, Cole-Parmer
Closures Safety Caps and Safety Waste Caps
The Vaplock™ closed system design reduces chemical evaporation and exposure to hazardous vapors.
  • Minimize exposure to harmful chemical vapors
  • Collect solvent waste from multiple effluent streams
  • Highly adaptable and conformable to a broad range of containment types
Featuring a vapor tight connection, waste manifolds are designed to fit 2" fine thread drums. Chemical-resistant materials of construction include polypropylene or PTFE manifold and insert, with polypropylene spin collar. These versatile manifolds can be configured for barbed connections,More Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable