Vaplock™ Justrite Container Quick Disconnect Waste Caps and Waste Kits, Cole-Parmer
Supplier: Antyila Scientific
12019-60 12019-58 12019-59 12019-52 12019-53 12019-54 12019-55 12019-56 12019-57
76287-462EA 425.33 CAD
76287-462 76287-864 76287-866 76287-970 76287-972 76287-974 76287-976 76287-978 76287-980
Vaplock™ Justrite Container Quick Disconnect Waste Caps and Waste Kits, Cole-Parmer
Closures Safety Caps and Safety Waste Caps
The Vaplock™ closed system design reduces chemical evaporation and exposure to hazardous vapors.
  • Minimize exposure to harmful chemical vapors
  • Collect solvent waste from multiple effluent streams
  • Highly adaptable and conformable to a broad range of containment types
  • Waste manifolds are designed to fit Justrite HPLC quick disconnect safety containers
Featuring a vapor tight connection, waste manifolds are designed to fit Justrite Container quick disconnect systems. Chemical-resistant materials of construction include polypropylene or PTFE manifold andMore Product Information