Vaplock™ Nalgene® Waste Caps and Waste Kits, 83B, Cole-Parmer
Supplier: Antyila Scientific
12019-44 12019-45 12019-40 12019-41 12019-42 12019-43
76287-558EA 776.8 CAD
76287-558 76287-560 76287-962 76287-964 76287-966 76287-968
Vaplock™ Nalgene® Waste Caps and Waste Kits, 83B, Cole-Parmer
Closures Safety Caps and Safety Waste Caps
The Vaplock™ closed system design reduces chemical evaporation and exposure to hazardous vapors.
  • Minimize exposure to harmful chemical vapors
  • Collect solvent waste from multiple effluent streams
  • Highly adaptable and conformable to a broad range of containment types
Featuring a vapor tight connection, waste manifolds are designed to fit Nalgene® 83B lab bottles. Chemical-resistant materials of construction include polypropylene or PTFE manifold and insert, with polypropylene spin collar. These versatile manifolds can be configured for barbedMore Product Information