Airfree® Schlenk Stoppers, O-Ring, Chemglass
Supplier: Chemglass
AF-0210-01 AF-0210-02
80064-394EA 67.84 CAD
80064-394 80064-396
Airfree® Schlenk Stoppers, O-Ring, Chemglass
Stoppers Glass Stoppers
For use with the Double Bank, Inert Gas or Single Bank, Inert Gas manifolds or any Airfree® component with an O-Ring as a closure.
Delivery information: Supplied complete with one Viton O-Ring.