Vaplock™ Nalgene® Supply Caps, 38-430, Cole-Parmer
Supplier: Antyila Scientific
12018-24 12018-25 12018-26 12018-27 12018-28 12018-29 12018-30 12018-31 12018-32 12018-33 12018-34 12018-35
76287-518EA 76.22 CAD
76287-518 76287-520 76287-522 76287-524 76287-526 76287-528 76287-530 76287-532 76287-534 76287-536 76287-538 76287-540
Vaplock™ Nalgene® Supply Caps, 38-430, Cole-Parmer
Closures Safety Caps and Safety Waste Caps
The Vaplock™ closed system design reduces chemical evaporation and exposure to hazardous vapors with Vaplock™ solvent reservoir caps.
  • Anti-Twist design prevents twisting of tubing when the cap is attached or removed, as the inner body rotates independently of the outer collar
  • Chemical compatibility with laboratory solvents
Durable port threads are designed to accept industry-standard 1/4-28 threaded fittings. Some caps include robust stainless steel thread inserts to minimize cross-threading and damage to soft PTFE ports. For use with semi-rigidMore Product Information