Airfree® Schlenk Connecting Adapters, Inlet, Chemglass
Supplier: Chemglass
AF-0074-01 AF-0074-02 AF-0074-03
80064-262EA 420.58 CAD
80064-262 80064-264 80064-266
Airfree® Schlenk Connecting Adapters, Inlet, Chemglass
Adapters Adapters, Joints with Tube Connectors
Connecting inlet adapter.
  • Equipped with two #15 O-Ring joints
  • Equipped with a 1-Arm Chem-Vac™ Chem-Cap® valve
Delivery information: Each adapter is supplied complete with two #116 Viton O-Ring.
Order Now
SPECIFICATIONS
Environmentally Preferable