Vaplock™ GL 38 Supply Caps, Cole-Parmer
Supplier: Antyila Scientific
12018-18 12018-21 12018-20 12018-23 12018-12 12018-13 12018-14 12018-15 12018-16 12018-17 12018-19 12018-22
76287-510EA 144.78 CAD
76287-510 76287-512 76287-514 76287-516 76287-716 76287-718 76287-720 76287-722 76287-724 76287-726 76287-942 76287-944
Vaplock™ GL 38 Supply Caps, Cole-Parmer
Closures Safety Caps and Safety Waste Caps
The Vaplock™ closed system design reduces chemical evaporation and exposure to hazardous vapors with Vaplock™ closed system solvent reservoir caps.
  • Anti-twist design prevents twisting of tubing when the cap is attached or removed as the inner body rotates independently of the outer collar
  • Chemical compatibility with laboratory solvents
Durable port threads are designed to accept industry-standard 1/4-28 threaded fittings. Some caps include robust stainless steel thread inserts to minimize cross-threading and damage to soft PTFE ports. For use with semi-rigid tubing equal to or less than 1/8" (3.2 mm) O.D.
Chemically compatible caps are constructed of inert PTFE. The body of the cap provides ideal chemical compatibility with laboratory solvents. Under normal use, the outer PP collar is non-wetted. Caps may be autoclaved to 140 °C. Combine with secure fluid connections and Vaplock air inlet or relief valves to maintain pressure equilibrium within the bottle.