Multiple Affinity Removal Spin Cartridge Reagent Kit, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES (GENOMICS) CA
5188-5254 5188-5252 5188-5251 5188-5250 5188-5249
CA76204-900EA 957.65 CAD
CA76204-900 CA76204-902 CA76204-904 CA76204-906 CA76204-908 CA76204-910
Multiple Affinity Removal Spin Cartridge Reagent Kit, Agilent Technologies
Protein Purification Protein Purification Kits
The affinity removal spin cartridge reagent kit is designed to remove the most interfering high-abundant protein (albumin) from human samples (such as plasma, serum, urine or cerebrospinal fluid).
Delivery information: Kit includes Buffer A, 1 bottle (1 l) Buffer B, 1 bottle (1 l), Spin filters, 2 packs, Spin concentrators, 5 KDa MWCo (4 ml) 1 pack, Luer-Lok adapter 1 pack, Luer-Lok syringes (5 ml) 1 pack, Microtubes, 6 packs, Caps and plugs 1 pack, PTFE Luer-Lok needles 1 pack.