Airfree® Cajon® Flexible Tubing, Schlenk, Chemglass
Supplier: Chemglass
AF-0069-09 AF-0069-01 AF-0069-02 AF-0069-03 AF-0069-04 AF-0069-05 AF-0069-06 AF-0069-08
80060-048EA 797.21 CAD
80060-048 80060-034 80060-036 80060-038 80060-040 80060-042 80060-044 80060-046
Airfree® Cajon® Flexible Tubing, Schlenk, Chemglass
Tubing Transfer Tubing
Cajon® Stainless Steel #321 Flexible Tubing is compressible by at least 20%, and extendable by 50% of the nominal listed length.
  • The ¹/₄" and ³/₈" sizes have a cuff length of 0.75"
  • The 1" size cuff length is 1"
Flexible tubing will not contract when evacuated, therefore glass apparatus should not be firmly clamped in place until vacuum is established within the system.