Vaplock™ GL 45 Supply Caps, Cole-Parmer
Supplier: Antyila Scientific
12018-01 12018-02 12018-03 12018-04 12018-05 12018-06 12018-07 12018-08 12018-09 12018-00 12018-10 12018-11
76287-496EA 119.52 CAD
76287-496 76287-498 76287-500 76287-502 76287-504 76287-506 76287-508 76287-652 76287-654 76287-678 76287-712 76287-714
Vaplock™ GL 45 Supply Caps, Cole-Parmer
Closures Safety Caps and Safety Waste Caps
The Vaplock™ closed system design reduces chemical evaporation and exposure to hazardous vapors with Vaplock™ closed system solvent reservoir caps.
  • Anti-twist design prevents twisting of tubing when the cap is attached or removed, as the inner body rotates independently of the outer collar
  • Chemical compatibility with laboratory solvents
Durable port threads are designed to accept industry-standard 1/4-28 threaded fittings. Some caps include robust stainless steel thread inserts to minimize cross-threading and damage to soft PTFE ports. For use withMore Product Information