Airfree® Schlenk Cajon® Cuffs, Chemglass
Supplier: Chemglass
AF-0068-01 AF-0068-02 AF-0068-03
80060-028EA 24.92 CAD
80060-028 80060-030 80060-032
Airfree® Schlenk Cajon® Cuffs, Chemglass
Tubing Transfer Tubing
Cuff adapters that do not require welding or brazing to tubing to insure a leak-tight seal.
  • Made of #304 stainless steel
  • MUST be used between unions and flexible tubing
  • Leak-tight seal