Vaplock™ Barbed Tubing Connections, Cole-Parmer
Supplier: Antyila Scientific
12020-18 12020-20 12020-21 12020-22 12020-23 12020-32 12020-24 12020-26 12020-28 12020-29 12020-30 12020-31 12020-34 12020-35
76287-600PK 39.42 CAD
76287-600 76287-602 76287-604 76287-606 76287-608 76287-610 76287-612 76287-614 76287-876 76287-878 76287-880 76287-882 76287-884 76287-886 76287-888 76287-890 76287-918 76287-920 76287-922
Vaplock™ Barbed Tubing Connections, Cole-Parmer
Fittings and Connectors
Barbed fittings are an economical means of connecting soft-wall tubing to manifold ports.
A semi-permanent connection for soft-walled tubing only, barbed fittings are an economical means of connecting tubing to Vaplock™ manifold ports with NPT threads.
Wrap NPT threads 3 to 4 times with PTFE tape and wrench tighten for leak-resistant connections.
For 1/16 to 1/2" I.D. tubing. All fittings are polypropylene. Select based on tubing inner diameter.