Airfree® Schlenk Drying Towers, KOH, for Portable Vacuum Systems, Chemglass
Supplier: Chemglass
AF-0300-50 AF-0300-51
80064-420EA 2401.04 CAD
80064-420 80064-422
Airfree® Schlenk Drying Towers, KOH, for Portable Vacuum Systems, Chemglass
Vacuum Traps
Drying tower is designed to fit on a cart system between the vacuum and pump oil trap.
  • Extra coarse fritted disc
  • 50 I.D×250 mm effective length
Trap has a sealed-in extra coarse fritted disc for support of KOH. Both ends have #20 O-Ring joints. Lower adapter has a #20 O-Ring joint with a 1" O.D. tubulation for connection to flexible vacuum tubing.
Delivery information: Complete item consists of one KOH tower, one lower tubing adapter, one 90°connection adapter, two #35 stainless steel pinch clamps, and two #214 Viton O-Rings.