Airfree® Schlenk Storage Vessels, Cajon®, Chemglass

Supplier: Chemglass
AF-0096-04 AF-0096-01 AF-0096-02 AF-0096-03
80064-350EA 560.9 CAD
80064-350 80064-344 80064-346 80064-348
Airfree® Schlenk Storage Vessels, Cajon®, Chemglass
Reaction Vessels Reaction Vessels, Flask Shape
Storage vessel with a Chem-Vac™ Chem-Cap® valve used to control gas or liquid flow into the vessel.

  • Equipped with either 0 to 4 mm or 0 to 8 mm Chem-Vac™, Chem-Cap® valve
  • For replacement valves, see (17003-148;17003-150)

Storage vessel sidearm has a 9 mm O.D. tubulation for use with 3/8" Cajon® connections. Chem-Vac™ Chem-Cap® valve is used to control gas or liquid flow into the vessel.
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR