Airfree® Schlenk Storage Vessels, with O-Ring, Chemglass

Supplier: Chemglass
AF-0092-01 AF-0092-02 AF-0092-03 AF-0092-04
80064-316EA 413.56 CAD
80064-316 80064-318 80064-320 80064-322
Airfree® Schlenk Storage Vessels, with O-Ring, Chemglass
Reaction Vessels Reaction Vessels, Flask Shape
Storage vessel complete with an O-Ring joint on the sidearm.

  • #15 O-Ring joint
  • Chem-Vac™, Chem-Cap® valve to control gas or liquid flow into the vessel

Vessels are constructed of medium wall tubing and are used with manifolds or any other Airfree® component with a #15 O-Ring joint.

Delivery information: Supplied complete with one #116 Viton O-Ring.
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR