Airfree® Schlenk Vacuum Manifolds, Single Bank, Inert Gas, Chemglass
Supplier: Chemglass
AF-0061-01 AF-0061-02 AF-0061-03
80064-222EA 1552.68 CAD
80064-222 80064-224 80064-226
Airfree® Schlenk Vacuum Manifolds, Single Bank, Inert Gas, Chemglass
Manifolds
Single bank vacuum manifold with 0 to 8 mm Chem-Vac™, Chem-Cap® valves mounted on a 45° angle to the user, permitting easy operation inside of a fume hood.
  • Ports are spaced 75 mm center to center
  • All connection ports are #15 O-Ring connections that incorporate a greasefree O-Ring seal
Manifolds are modular in design, allowing the user tremendous flexibility in performing experiments. Adapters from #15 O-Ring joints to standard taper components, serrated hose connections, or 3/8" Cajon® fittings are available to allow the ports to be used with existing glassware.
For replacement Chem-Vac™ Chem-Cap® plugs, see (17003-150).