Airfree® Schlenk Horizontal Bubblers, Chemglass
Supplier: Chemglass
AF-0514-01 AF-0514-02
80064-570EA 572.56 CAD
80064-570 80064-572
Airfree® Schlenk Horizontal Bubblers, Chemglass
Bubblers
Horizontal bubblers manufactured by Chemglass.
  • Allows for in-line connection
  • With 9 mm O.D. middle serration
  • Equipped with standard taper inner joint
Horizontal design of this bubbler allows for in-line connection and purging of the apparatus. Hose connection has a 9 mm O.D. middle serration. Complete with standard taper inner joint.
Order Now
SPECIFICATIONS
Environmentally Preferable