Airfree® Schlenk Flushing Adapters, without Septum Stoppers, Chemglass
Supplier: Chemglass
AF-0509-A-01 AF-0509-A-02 AF-0509-A-03 AF-0509-A-04
80064-538EA 427.29 CAD
80064-538 80064-540 80064-542 80064-544
Airfree® Schlenk Flushing Adapters, without Septum Stoppers, Chemglass
Adapters Adapters, Conical Ground Joints
Similar to flushing adapter but having a top outer joint and lower inner joint of the same size.
  • Stopcocks have 9 mm O.D. medium wall arms
  • Addition of the outer joint permits use of the larger sizes septum stoppers for repeated needle penetrations
Adapter also allows for evacuation and transfer of product into another vessel with the same size standard taper joints.
Ordering information: These adapters are not supplied with septum stopper.
Delivery information: Available with a choice of a 4 mm glass high vacuum stopcock with a stainless steel constant pressure retention device or 4 mm PTFE stopcock.