Airfree® Schlenk Flushing Adapters, with Septum Stoppers, Chemglass
Supplier: Chemglass
AF-0508-10 AF-0508-11 AF-0508-20 AF-0508-21
80064-518EA 363.12 CAD
80064-518 80064-520 80064-522 80064-524
Airfree® Schlenk Flushing Adapters, with Septum Stoppers, Chemglass
Adapters Adapters, Conical Ground Joints
Adapter permits vessels to be purged with an inert gas during sampling or transfer with a cannulas.
  • Stopcocks have 9 mm O.D. medium wall arms
Both the glass and PTFE stopcock have 9 mm O.D. medium wall arms for use with red or white septum stopper. Additional arm is provided for connection to inert gas source.
Delivery information: Available with choice of 4 mm high vacuum glass stopcock or 4 mm PTFE stopcock. Supplied with two red septum stoppers.