Airfree® Schlenk Connecting Adapters, Styles A and B, Chemglass
Supplier: Chemglass
AF-0506-01 AF-0506-02 AF-0506-03 AF-0506-04
80064-502EA 499.92 CAD
80064-502 80064-504 80064-506 80064-508
Airfree® Schlenk Connecting Adapters, Styles A and B, Chemglass
Adapters Adapters, Conical Ground Joints
Connecting adapters available in two different styles to best suit your needs.
  • Available in two configurations
  • Style A features the valve mounted in-line
  • Style B features the valve 90° to joints with a brace permitting hanging of heavier objects from the line
Adapters have a 0 to 4 mm Chem-Vac™ Chem-Cap® valve between the top outer and lower inner joints, both of which are the same size. Overall length of the 14/20 size is approximately 135 mm, while the 24/40 length is approximately 190 mm.
For replacement Chem-Vac™ Chem-Cap® valve, see 17003-148.