Airfree® Schlenk 2-Neck Reaction Flasks, Chemglass
Supplier: Chemglass
AF-0530-01 AF-0530-02 AF-0530-03 AF-0530-04 AF-0530-05 AF-0530-06 AF-0530-20 AF-0530-21 AF-0530-22 AF-0530-23 AF-0530-24 AF-0530-25
80064-690EA 423.22 CAD
80064-690 80064-692 80064-694 80064-696 80064-698 80064-700 80064-702 80064-704 80064-706 80064-708 80064-710 80064-712
Airfree® Schlenk 2-Neck Reaction Flasks, Chemglass
Reaction Vessels Reaction Vessels, Flask Shape
Round bottom, two neck flasks have standard taper outer joints, both being the same size.
  • Equipped with either a 2 mm high vacuum glass stopcock or a 0 to 4 mm Chem-Cap® Airfree® inlet valve
  • Available in 14/20 or 24/40 joint sizes
Available with a choice of a 2 mm high vacuum glass stopcock with a stainless steel constant pressure retaining device, or a 0 to 4 mm Chem-Cap® Airfree® inlet valve. Side neck can be fitted with septum stopper for sampling or cannula transfer.