SP Bel-Art H-B® DURAC® Alcohol Proof, Ethyl Alcohol Hydrometers, Traceable to NIST, Bel-Art Products, a part of SP
Supplier: Bel-Art Products, a Part of SP
B61807-4300 B61807-4400
76287-330CS 2368.93 CAD
76287-330 76287-332
SP Bel-Art H-B® DURAC® Alcohol Proof, Ethyl Alcohol Hydrometers, Traceable to NIST, Bel-Art Products, a part of SP
Hydrometers
Used to determine proof of ethyl alcohol spirits, these hydrometers are available in various ranges.
  • 300 mm (12") long
  • Lead free ballast and glass
  • Individually serialized
  • Temperature standardization: 60 °F (15 °C)
  • Hand inspected by our U.S. quality team
Certifications: Made according to the Bureau of Internal Revenue, U.S. Treasury Department, the tolerances listed for these hydrometers are those outlined in Circular 555 of the National Bureau of Standards, Testing of Hydrometers.
Packaging: Packaged with a multi-language (English, French, German, Italian, Portuguese, and Spanish) Statement of Accuracy indicating accuracy traceable to NIST.
Caution: Hydrometers should be stored in a clean, dry, safe place and should not be subjected to extreme temperatures.