Brilliant III Ultra-Fast Probe ROX qPCR Master Mix, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES (GENOMICS) CA
600890 600888 600899 600898
CA76204-814EA 457.35 CAD
CA76204-814 CA76204-818 CA76204-852 CA76204-854
Brilliant III Ultra-Fast Probe ROX qPCR Master Mix, Agilent Technologies
Nucleic Acid Reagents qPCR/RT-qPCR Enzymes and Kits
High performance, ultra-sensitive, probe QPCR master mix reagent with high or low ROX concentration for reliable quantification across a wide range of targets and templates.
  • Resistant to common QPCR inhibitors (whole blood, humic acid, NaCl)
  • Extremely fast while maintaining sensitivity
  • Optimized fast cycling formulation ensures reliable and reproducible data with shorter run times
Brilliant III Ultra-Fast maintains amplification efficiency, R2, dynamic range and detection sensitivity all while shortening the total cycling time by ~60% compared to standard cycling.
High performance, ultra-sensitive, probe QPCR master mix reagent with high/low ROX concentration for reliable quantification across a wide range of targets and templates. Convenient pre-blended formulations compatible with any sequence-specific probe detection chemistry.
Caution: For research use only. Not for use in diagnostic procedures.
Order Now
SPECIFICATIONS
Environmentally Preferable
Application qPCR probe