HyClone™ HyCell CHO Cell Culture Media, HyClone products (Cytiva)
Supplier: Cytiva
CA76237-080EA 478.21 CAD
CA76237-080 CA76237-082 CA76237-084
HyClone™ HyCell CHO Cell Culture Media, HyClone products (Cytiva)
Cell Culture Media
HyCell™ CHO media has been developed for CHO cell culture for bioprocess applications.
  • Animal-derived component free (ADCF)
  • Allows for direct or sequential adaptation
  • Component traceability
  • Manufactured under cGMP
This media has been specifically formulated using Metabolic Pathway Design. It provides optimal performance and supports higher peak cell density, enhanced product yield, and quality. The versatility of HyCell™ CHO media allows quick adaptation and supports exceptional growth, high density, and high productivity up to five-fold or more over other comparable media across a broad variety of CHO clones.