HyClone™ Classical Liquid Media, Dulbeccos Modified Eagles Medium, Low Glucose, HyClone products (Cytiva)
Supplier: Cytiva
SH30021.01 SH30021.FS
CA16777-124EA 36.86 CAD
CA16777-124 CA16777-127 CA16750-094
HyClone™ Classical Liquid Media, Dulbeccos Modified Eagles Medium, Low Glucose, HyClone products (Cytiva)
Cell Culture Media
DMEM Low Glucose was the original modification of Basal Medium Eagle (BME) used to grow mouse embryonic cells. DMEM, Low Glucose is suitable for the culturing of many immortalised cell lines from humans, monkeys, hamsters, rats, mice, chicken and fish. DMEM contains four times more amino acids, vitamins and other supplementary components than Basal Medium Eagle (BME), the base media from which DMEM was derived. As DMEM contains no growth-promoting agents or proteins, supplementation with FBS is recommended for optimal performance.
  • Contains 5,6 mM glucose
  • Produced with WFI quality water
  • 0.1 μm sterile filtered
  • Manufactured using ISO 9001:2000 certified processes