CHEMets® Visual Kit, Hydrogen Peroxide Test Kit, CHEMetrics
Supplier: CHEMetrics
K-5510 R-5510
CXK5510EA 265.68 CAD
CXK5510 CXR-5510
CHEMets® Visual Kit, Hydrogen Peroxide Test Kit, CHEMetrics
Environmental Test Kits
Hydrogen Peroxide CHEMets® used to to monitor sterilisation solutions in the packaging and sanitizing processes.
Hydrogen peroxide is a strong oxidizing agent used for treating of industrial effluents and domestic waste and serving as a disinfectant in aseptic packaging.
Delivery information: Kit includes refill, low and high range comparators, 25 ml sample cup, and instructions.
Packaging: Comes in a plastic case.