Triphenylphosphine ≥99.5% (by GC), crystals
Supplier: CHEM-IMPEX INTERNATIONAL, INC
02365-100G
02365-1KG
02365-250G
CH02365-100GEA
29.28
CAD
CH02365-100G
CH02365-1KG
CH02365-250G
Triphenylphosphine ≥99.5% (by GC), crystals
Triphenylphosphine
Used in the synthesis of organic and organometallic compounds.
Formula:
C₁₈H₁₅P MW: 262.29 g/mol Boiling Pt: 377 Melting Pt: 79 Flash Pt: 180 Storage Temperature: Ambient |
MDL Number:
MFCD00003043 CAS Number: 603-35-0 Merck Index: 12,09873 |
Specification Test Results
Purity | ≥99.5% (GC) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...