Injection Training Pads
Supplier: WALLCUR, LLC.
REM-INJ-SP-01L REM-INJ-LG-01L REM-INJ-EPI-01
470306-768EA 57.19 CAD
470306-768 470306-770 470306-772
Injection Training Pads
Models Human Biology Models
Remedy's medical training injection pads are designed for the practice of needle injections.
The pads can be used to simulate intradermal, intra-muscular and subcutaneous injections. The larger pad allows you to have more practice area, as well as practicing Z-track injections. A removable epidermis layer is placed on the surface of the pad allowing for the simulation of a "wheal" during intradermal injections.
The skin puncture resistance, as well as the resistance throughout the injection, closely mimics the feel of a real injection. Both pads have drainage on theMore Product Information