Commander™ Safety Glasses, Protective Industrial Products
Supplier: PIP
250-33-0020 250-33-0226 250-33-1020
10120-249CS 5885.23 CAD
10120-249 10120-251 10120-253
Commander™ Safety Glasses, Protective Industrial Products
Safety Glasses
Lightweight and semi-rimless design offers eye coverage and comfort.
  • Bayonet-style temples with non-slip rubber pads provide a secure fit
  • Adjustable rubber nose pads
  • FogLess™ lens coating as standard
The 9.5 base curve lens are used with anti fog and anti scratch coating.
Certifications: Meets ANSI Z87.1 and CSA Z94.3 requirements.
Order Now
SPECIFICATIONS
Environmentally Preferable