Gravity convection laboratory ovens are recommended for critical scientific and analytical use in laboratory and pilot plant product research. Typical applications include aging, curing, drying, baking, pre-heating, conditioning and life cycle testing.
- Fast ramp-up to 306 °C for faster cycle times
- Temperature range approximately 20 °C above ambient to 306 °C for multiple applications
- Advanced PID temperature control system for sensitive response
- Built-in digital timer for automatic operation following manual start
- Independent over temperature set-point and operational control override for additional safety
- Interior construction with 430 series stainless steel
- Stainless steel shelves are adjustable on 0.5" (12.7 mm) centers to provide flexible inventory options
SGO Series ovens are available in 110 to 120 V, 220 to 240 V, 50/60 Hz configurations. These feature forced-air circulation to assure maximum temperature uniformity, triple-wall construction to minimize external surface temperature.
Rear access port, 1.75" diameter (44 mm) for independent cables, instrumentation, Exhaust port, 1.75" diameter (44 mm) adjustable, with external connection to customer-supplied vent for effluent exhaust if required.
Upper vents are closed when recovery times following door openings are tested. Published recovery performance is based on a standard cabinet under controlled testing at a (22 ± 3 °C ambient temperature, line voltage ±10% of specified voltage.
Certifications: Internationally certified CAN/CSA, UL, EN, IEC 61010, and compliant with CE.