Antistatic Polystyrene Hexagonal Weighing Dishes, Simport Scientific
Supplier: Simport Scientific
D252-2 D252-3 D252-4 D252-1 D252-1 D252-4
CA11016-304CS 69.3 CAD
CA11016-304 CA11016-306 CA11016-308 CA10798-880 76299-242 76299-244
Antistatic Polystyrene Hexagonal Weighing Dishes, Simport Scientific
Boats Weighing Boats
Ideal for handling solids or liquids during weighing.
  • Easily bent into pouring spouts
  • Dishes enable non-spill transfer
  • Stackable for easy storage
  • Withstands temperatures up to 80 °C
Molded hexagonal design provides greater balance protection and safety. Will not react with most substances. Suitable for weighing static-affected samples.