Cloning Cylinders, Glass, Chemglass Inc.
Supplier: Chemglass
CLS-1777-03 CLS-1777-01 CLS-1777-02
10170-798PK 198.58 CAD
10170-798 10170-800 10170-802
Cloning Cylinders, Glass, Chemglass Inc.
Cylinders Cloning Cylinders
Clone a group or single cell by surrounding them with these glass cylinders.
  • Autoclavable or can be dry heat sterilized
The cylinders are sealed to the plate with sterile silicone high vacuum grease, such as Dow Corning 976v thereby forming an isolation well.
Delivery information: Supplied in package of 125 pieces.