Polyethyleneimine, M.W. 40,000, linear
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Polyaziridine, Corcat, Polyethylenimine, Polymin P, Polyaziridine Mw 25000, Aziridine, PEI, Polyethylenimine (branched), Ethyleneimine Polymer, Poly(ethyleneimine)
047336-03
047336-MD
AA47336-03EA
4709.49
CAD
AA47336-03
AA47336-MD
Polyethyleneimine, M.W. 40,000, linear
Polyethyleneimine
Formula:
(CH2CH2NH)n Density: 1.05 Storage Temperature: Ambient |
MDL Number:
MFCD00084427 CAS Number: 9002-98-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...