Calcined petroleum coke powder -20+80 mesh

Supplier: Thermo Scientific Chemicals

047287-30 047287-A1
AA47287-30EA 59.25 CAD
AA47287-30 AA47287-A1
Calcined petroleum coke powder -20+80 mesh
Calcined petroleum coke powder
Density: 2,1
Storage Temperature: Ambient
CAS Number: 64743-05-1
EINECS: 265-210-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR