Rosmarinic acid ≥97%

Supplier: Thermo Scientific Chemicals
Danger

H37451-03 H37451-06
AAH37451-03EA 170.64 CAD
AAH37451-03 AAH37451-06
Rosmarinic acid ≥97%
Rosmarinic acid
Formula: C₁₈H₁₆O₈
Melting Pt: 171…175 °C
Storage Temperature: Ambient
MDL Number: MFCD00017740
CAS Number: 20283-92-5
EINECS: 606-487-1
UN: 2811
ADR: "6.1",III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR