Oxymatrine

Supplier: Thermo Scientific Chemicals
Warning

H37340-03 H37340-MD
AAH37340-03EA 641.48 CAD
AAH37340-03 AAH37340-MD
Oxymatrine
Oxymatrine
Melting Pt: 208 °C
Storage Temperature: Ambient
MDL Number: MFCD00210339
CAS Number: 16837-52-8
EINECS: 605-514-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR